Learn More
Fine Science Tools Fine Scissors, CeramaCut, Curved, 11.5 cm
SDP
Catalog No. NC9495743
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Ceramic Coated Stainless Steel/ Length: 11.5cm/ Serrated: Yes/ Feature: CeramaCut
Catalog No. NC9495743 Supplier Fine Science Tools Supplier No. 1495911
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Ceramic Coated Stainless Steel/ Length: 11.5cm/ Serrated: Yes/ Feature: CeramaCut