Learn More
World Precision Instrument Ceramic-Coated Cupped Forceps, 12.5cm, Straight, 3.5mm Tip
SDP
Catalog No. 502521365
Encompass
Cupped Forceps have small (3.5mm), smooth, round cups at the tip, which work well in microdissection, small animal surgery, electronics repair, and some crafts like beading. The tips of these WPI cupped forceps are coated with either Teflon or ceramic, giving you a smooth, non-stick surface to reduce tissue trauma. These tips are low-glare, perfect when you are working under magnification or surgical lights. The coating makes the tips chemically inert, resistant to staining and easy to clean. They are also more resistant to wear. Teflon or ceramic coatings offer electrical insulation, which can be critical during electrosurgery. These cupped forceps are made is Switzerland of surgical grade stainless steel. They are suitable for sterilization via autoclave or chemical sterilant.
Catalog No. 50-252-1365 Supplier World Precision Instrument Supplier No. 501984
Cupped Forceps have small (3.5mm), smooth, round cups at the tip, which work well in microdissection, small animal surgery, electronics repair, and some crafts like beading. The tips of these WPI cupped forceps are coated with either Teflon or ceramic, giving you a smooth, non-stick surface to reduce tissue trauma. These tips are low-glare, perfect when you are working under magnification or surgical lights. The coating makes the tips chemically inert, resistant to staining and easy to clean. They are also more resistant to wear. Teflon or ceramic coatings offer electrical insulation, which can be critical during electrosurgery. These cupped forceps are made is Switzerland of surgical grade stainless steel. They are suitable for sterilization via autoclave or chemical sterilant.