Learn More
Fine Science Tools Ceramic Coated Spring Scissors, Straight, 10 cm
SDP
Catalog No. NC1575248
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 7mm/ Tip Diameter: 0.1mm/ Alloy / Material: Ceramic Coated Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC1575248 Supplier Fine Science Tools Supplier No. 1575011
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 7mm/ Tip Diameter: 0.1mm/ Alloy / Material: Ceramic Coated Stainless Steel/ Length: 10cm/ Serrated: No