Learn More
Chemglass Life Sciences AREC.X Digital Ceramic Hotplate Stirrer with PT100 Probe, 115V/60Hz
SDP
Catalog No. NC1292420
Encompass_Preferred
Ceramic Digital Hotplate Stirrer, 7.9 x 7.9", 115V, Complete with 3mm PT100 Probe. Compliance with the following standards CE, CSA and UL. Square hot plate offers advanced technology with a digital display for stirring speed and temperature settings. A microprocessor ensures constant speed even when viscosity changes. The 7.9x7.9¿ white ceramic hotplate surface is easy to clean and provides excellent resistance to chemicals, scratches and surface abrasions. Hot plate designed for precise thermoregulation as well as a high degree of reliability and safety. Designed for maximum protection against liquid penetration. Unit features an elevated front panel and runoff groove. Hotplate stirrer comes equipped with a socket for the connection of the optional VTF digital thermoregulator for more precise temperature control and a digital count down timer that shuts off heating- adjustable from 0 to 24:59 (h:min). Supplied complete with probe only. Rod, clamp purchased separately.
Catalog No. NC1292420 Supplier Chemglass Life Sciences Supplier No. CG1999X10
Ceramic Digital Hotplate Stirrer, 7.9 x 7.9", 115V, Complete with 3mm PT100 Probe. Compliance with the following standards CE, CSA and UL. Square hot plate offers advanced technology with a digital display for stirring speed and temperature settings. A microprocessor ensures constant speed even when viscosity changes. The 7.9x7.9¿ white ceramic hotplate surface is easy to clean and provides excellent resistance to chemicals, scratches and surface abrasions. Hot plate designed for precise thermoregulation as well as a high degree of reliability and safety. Designed for maximum protection against liquid penetration. Unit features an elevated front panel and runoff groove. Hotplate stirrer comes equipped with a socket for the connection of the optional VTF digital thermoregulator for more precise temperature control and a digital count down timer that shuts off heating- adjustable from 0 to 24:59 (h:min). Supplied complete with probe only. Rod, clamp purchased separately.