Learn More
RWD LIFE SCIENCE INC CERAMIC FERRULE HOLDER D1.25M
SDP
Supplier:  RWD LIFE SCIENCE INC 68214
Encompass
Hold cannula or Ceramic Ferrule during implantation. Extended double-leaf clamping design reduces visual field obstruction. Versions for Dia. 1.25 mm — 68214 Cannula or Ceramic Ferrule.
Catalog No. NC2480300