Learn More
Thermo Scientific™ Ceramic Homogenizers for QuEChERS Kits
Catalog No. 01184139 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
15 mL Tubes
2 mL Tubes
50 mL Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-184-139 2 mL Tubes
01-184-140 15 mL Tubes
01-184-141 50 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-184-139 Supplier Thermo Scientific™ Supplier No. 60106CH2
Add to Cart
Add to Cart
Use Thermo Scientific™ Ceramic Homogenizers to grind and homogenize samples for your QuEChERS sample preparation workflow.
Use Thermo Scientific™ Ceramic Homogenizers to help accelerate the homogenization process and increase recovery and reproducibility during your QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) sample preparation workflow. These homognizers can be used with all of our QuEChERS Extraction and Clean-Up kits to supplement Original 2003 Method, AOAC 2007.01 Method, EN 15662 Method, and NY/T 1380-2007 Method procedures.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 2 mL Tubes
Type Ceramic Homogenizer
Quantity 200/Pk.