Learn More
Glass Expansion Ceramic Injector 1.25mm Tapered
SDP
Catalog No. 501907260
Encompass_Preferred
Alumina Tapered Injector with 1.25mm ID. 1.25mm ID tapered quartz injector (N0791181). For use with Avio 200/220 and 500/550 ICP-OES series. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents such as xylene.Compatible with PerkinElmer Avio 200/220 D-Torch (P/N 50-190-7038), Avio 500/550 D-Torch (P/N 50-190-7039), Avio 200/220 Adjustable torch body (P/N 50-211-3618) and Avio 500/550 Adjustable torch body (P/N 50-211-2626).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7260 Supplier Glass Expansion Supplier No. 318083799
Alumina Tapered Injector with 1.25mm ID. 1.25mm ID tapered quartz injector (N0791181). For use with Avio 200/220 and 500/550 ICP-OES series. Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia. They are also used with less volatile organic solvents such as xylene.Compatible with PerkinElmer Avio 200/220 D-Torch (P/N 50-190-7038), Avio 500/550 D-Torch (P/N 50-190-7039), Avio 200/220 Adjustable torch body (P/N 50-211-3618) and Avio 500/550 Adjustable torch body (P/N 50-211-2626).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001