Learn More
Glass Expansion Ceramic Outer Tube for D-Torch, Optima 8x00
SDP
Supplier:  Glass Expansion 318082990
Encompass_Preferred
Ceramic Outer Tube for D-Torch. Compatible with PerkinElmer Optima 8000/8300 DV Series ICP-OES D-Torch (30-808-3300, 501907027). Compared to quartz outer tubes, the ceramic alternative has an extended lifespan, significantly reducing maintenance, cleaning, and downtime caused by torch failure. Incorporating a ceramic outer tube into your ICP torch has a range of benefits, particularly for the analysis of high-TDS sample matrices, fusions, wear metals in oil, hydrofluoric acid (HF) and volatile organics. In addition to providing durability, the ceramic outer tube on an ICP torch produces a hotter, more robust plasma, which reduces matrix effects and improves sensitivities and detection limits, compared to a quartz torch.When not in use, clean as per recommended guidelines, and store in original box.Optional quartz outer tube P/N 501907204 and 31-808-3407 (no slot).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7170