Learn More
Glass Expansion Ceramic Outer Tube & Optic Fiber for D-Torch
SDP
Catalog No. 502703146
Encompass_Preferred
Ceramic Outer Tube & Optic Fiber for D-Torch. Compatible with Thermo 6000/7000 Duo ICP-OES. Includes Ceramic Outer Tube (501694666, 31-808-3834), Optic Fiber 170mm (31-808-2933), and Optic Fiber 450mm (31-808-3117). The shorter optical fiber assembly is used for Thermo 6000 models up to the year 2010, while the longer optical fiber assembly is used from the year 2011 onwards.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3146 Supplier Glass Expansion Supplier No. 318083079
Ceramic Outer Tube & Optic Fiber for D-Torch. Compatible with Thermo 6000/7000 Duo ICP-OES. Includes Ceramic Outer Tube (501694666, 31-808-3834), Optic Fiber 170mm (31-808-2933), and Optic Fiber 450mm (31-808-3117). The shorter optical fiber assembly is used for Thermo 6000 models up to the year 2010, while the longer optical fiber assembly is used from the year 2011 onwards.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001