Learn More
TA Instruments Ceramic Sample Pans 100µL Pkg of 3, for Use with All Discovery TGAs, Temperature Range Ambient to 1200°C
Catalog No. 5000580629 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Ceramic Sample Pans 100 µL (pkg of 3) are designed for use with all Discovery TGA instruments. They can be used over the temperature range of ambient to 1200 degrees Celsius.
Catalog No. 50-005-80629 Supplier TA Instruments Supplier No. 957329.903
Waters TA Instruments Ceramic Sample Pans 100 µL (pkg of 3) are designed for use with all Discovery TGA instruments. They can be used over the temperature range of ambient to 1200 degrees Celsius.