Learn More
TA Instruments Ceramic Sample Pans 250µL Pkg of 3, for Use with All Discovery TGAs, Temperature Range Ambient to 1200°C
Catalog No. 5000580741 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Ceramic Sample Pans 250 µL (pkg of 3) are designed for use with all Discovery TGA instruments. They can be used over the temperature range of ambient to 1200 degrees Celsius.
Catalog No. 50-005-80741 Supplier TA Instruments Supplier No. 957329.904
Waters TA Instruments Ceramic Sample Pans 250 µL (pkg of 3) are designed for use with all Discovery TGA instruments. They can be used over the temperature range of ambient to 1200 degrees Celsius.