Learn More
Perkin Elmer US LLC Ceramic Sample Pans - Qty. 5
Catalog No. 502111370
Encompass_Preferred
These ceramic sample pans are designed to hold a variety of substances, both volatile and non-volatile, and to withstand temperatures ranging from sub-ambient to over 1000 °C.
Catalog No. 50-211-1370 Supplier Perkin Elmer US LLC Supplier No. N5320103
These ceramic sample pans are designed to hold a variety of substances, both volatile and non-volatile, and to withstand temperatures ranging from sub-ambient to over 1000 °C.