Learn More
Electron Microscopy Sciences EMS Ceramic Tweezers Style 8507, Curved Tips, 125 mm
SDP
Catalog No. 5023779
Encompass_Preferred
Zirconia Toughened Alumina (ZTA) is a advanced ceramic compound that has a superior combination of high strength (from zirconia) and extreme hardness (from alumina). The density is relatively low and there is no open porosity. It is characterized by a very hard surface with good abrasion and wear resistance and good flexural strength and fracture toughness.
Stable at high temperatures, corrosion resistance, nearly chemically inert, and electrically insulating, ZTA is ideal for soldering processes and handling of components during thermal and chemical processes. These tweezers are used when very rigid tips are required and are suited for working with acids (except HF), for SDM, solders pot, and high temperature exposure. Clean room approved. Can be sterilized.
OAL: 125 mm
Tips: Curved, tips 1 mm wide, smooth
Catalog No. 50-237-79 Supplier Electron Microscopy Sciences Supplier No. 78120
Zirconia Toughened Alumina (ZTA) is a advanced ceramic compound that has a superior combination of high strength (from zirconia) and extreme hardness (from alumina). The density is relatively low and there is no open porosity. It is characterized by a very hard surface with good abrasion and wear resistance and good flexural strength and fracture toughness.
Stable at high temperatures, corrosion resistance, nearly chemically inert, and electrically insulating, ZTA is ideal for soldering processes and handling of components during thermal and chemical processes. These tweezers are used when very rigid tips are required and are suited for working with acids (except HF), for SDM, solders pot, and high temperature exposure. Clean room approved. Can be sterilized.
OAL: 125 mm
Tips: Curved, tips 1 mm wide, smooth