Learn More
Medchemexpress LLC Ceramide 3 | 34354-88-6 | >98.0% | C36H73NO4 | 1 MG
SDP
Catalog No. 5000356821
Encompass_Preferred
Ceramide 3 (also known as N-Stearoyl phytosphingosine or C18 Phytoceramide) is a bioactive sphingolipid naturally occurring in the stratum corneum of organisms like Saccharomyces cerevisiae, wheat grain, and mammalian epidermis. It is structurally composed of a phytosphingosine backbone amine-linked to a C18 fatty acid chain.
  • Regulates apoptosis, cell differentiation, and proliferation of smooth muscle cells
  • Inhibits the mitochondrial respiratory chain
  • Suppresses the expression of allergic cytokines IL-4, TNF-α, and transcription factors c-Jun and NF-κB in histone-stimulated mouse skin tissue
  • Formulations are used as skin protectants in cosmetics
  • Helps reduce water loss and prevent epidermal dehydration and irritation
Catalog No. 50-003-56821 Supplier Medchemexpress LLC Supplier No. HY1415821MG
Ceramide 3 (also known as N-Stearoyl phytosphingosine or C18 Phytoceramide) is a bioactive sphingolipid naturally occurring in the stratum corneum of organisms like Saccharomyces cerevisiae, wheat grain, and mammalian epidermis. It is structurally composed of a phytosphingosine backbone amine-linked to a C18 fatty acid chain.
  • Regulates apoptosis, cell differentiation, and proliferation of smooth muscle cells
  • Inhibits the mitochondrial respiratory chain
  • Suppresses the expression of allergic cytokines IL-4, TNF-α, and transcription factors c-Jun and NF-κB in histone-stimulated mouse skin tissue
  • Formulations are used as skin protectants in cosmetics
  • Helps reduce water loss and prevent epidermal dehydration and irritation