Learn More
Medchemexpress LLC Ceramide 3B | 178436-06-1 | 99.6% | 581.95 | 1 ML
SDP
Catalog No. 5000393139
Encompass_Preferred
Ceramide 3B is a ceramide 3 lipid class characterized by its aggregation behavior. It possesses various physiological functions, including acting as a penetration enhancer and an auxiliary emulsifier, and is significant for maintaining barrier function stability.
  • Lipid class: Ceramide 3 lipid class
  • Exhibits aggregation behavior
  • Has a variety of physiological functions
  • Can serve as a penetration enhancer
  • Functions as an auxiliary emulsifier
  • Plays an important role in barrier function stability
Catalog No. 50-003-93139 Supplier Medchemexpress LLC Supplier No. HY16041010MM/1ML
Ceramide 3B is a ceramide 3 lipid class characterized by its aggregation behavior. It possesses various physiological functions, including acting as a penetration enhancer and an auxiliary emulsifier, and is significant for maintaining barrier function stability.
  • Lipid class: Ceramide 3 lipid class
  • Exhibits aggregation behavior
  • Has a variety of physiological functions
  • Can serve as a penetration enhancer
  • Functions as an auxiliary emulsifier
  • Plays an important role in barrier function stability