Learn More
Medchemexpress LLC Ceramide 3B | 178436-06-1 | 99.6% | 581.95 | 100 MG
SDP
Catalog No. 5000391692
Encompass_Preferred
Ceramide 3B is a ceramide 3 lipid class characterized by its aggregation behavior. It possesses various physiological functions, including acting as a penetration enhancer and an auxiliary emulsifier, contributing significantly to barrier function stability.
  • Has aggregation behavior
  • Can serve as a penetration enhancer
  • Can serve as an auxiliary emulsifier
  • Plays an important role in barrier function stability
Catalog No. 50-003-91692 Supplier Medchemexpress LLC Supplier No. HY160410100MG
Ceramide 3B is a ceramide 3 lipid class characterized by its aggregation behavior. It possesses various physiological functions, including acting as a penetration enhancer and an auxiliary emulsifier, contributing significantly to barrier function stability.
  • Has aggregation behavior
  • Can serve as a penetration enhancer
  • Can serve as an auxiliary emulsifier
  • Plays an important role in barrier function stability