Learn More
Medchemexpress LLC Ceramide 3B | 178436-06-1 | 99.6% | 581.95 | 25 MG
SDP
Catalog No. 5000394308
Encompass_Preferred
Ceramide 3B is a ceramide 3 lipid class that exhibits aggregation behavior. It possesses various physiological functions, including acting as a penetration enhancer and an auxiliary emulsifier, and plays a significant role in maintaining barrier function stability.
  • Functions as a penetration enhancer.
  • Acts as an auxiliary emulsifier.
  • Contributes to barrier function stability.
Catalog No. 50-003-94308 Supplier Medchemexpress LLC Supplier No. 25MGHY160410
Ceramide 3B is a ceramide 3 lipid class that exhibits aggregation behavior. It possesses various physiological functions, including acting as a penetration enhancer and an auxiliary emulsifier, and plays a significant role in maintaining barrier function stability.
  • Functions as a penetration enhancer.
  • Acts as an auxiliary emulsifier.
  • Contributes to barrier function stability.