Learn More
Apexbio Technology LLC Ceramide(Synonyms: N-Acylsphingosine, Ceramides, Acylsphingosine, sphingosine N-acyl derivative, N-Acyl-D-sphingosine), 5mg, CAS: 3102-57-6.
SDP
Catalog No. 5000060495
Encompass_Preferred
Ceramide (CAS 3102-57-6) is a bioactive lipid molecule known to modulate diverse cellular processes including proliferation and differentiation It exerts its effects primarily by activating several serine/threonine protein phosphatases notably protein phosphatase-1 (PP1) protein phosphatase-2A (PP2A) and ceramide-activated protein phosphatase (CAPP) in vitro Due to its regulatory role in fundamental signaling pathways ceramide serves as a valuable research tool in investigations addressing cell cycle progression apoptosis and cellular homeostasis
Catalog No. 50-000-60495 Supplier Apexbio Technology LLC Supplier No. A45345
Ceramide (CAS 3102-57-6) is a bioactive lipid molecule known to modulate diverse cellular processes including proliferation and differentiation It exerts its effects primarily by activating several serine/threonine protein phosphatases notably protein phosphatase-1 (PP1) protein phosphatase-2A (PP2A) and ceramide-activated protein phosphatase (CAPP) in vitro Due to its regulatory role in fundamental signaling pathways ceramide serves as a valuable research tool in investigations addressing cell cycle progression apoptosis and cellular homeostasis