Learn More
Echelon Biosciences Research Labs CERAMIDE BEADS 1 mL
SDP
Catalog No. 50720694
Encompass
Ceramide Beads are composed of agarose with 10 nanomoles of bound ceramide per mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-694 Supplier Echelon Biosciences Research Labs Supplier No. PBCER1ML
Ceramide Beads are composed of agarose with 10 nanomoles of bound ceramide per mL of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable