Learn More
Cayman Chemical Ceramide Phosphoethanolamine (bovine), Purity: ≥98%, 5 mg
SDP
Catalog No. 502934137
Encompass_Preferred
A mixture of ceramide phosphoethanolamines; produced and used by invertebrates such as Drosophila, which lacks sphingomyelin and produced only in very small amounts in mammals; has a role in glial ensheathment of axons in Drosophila; depletion in vitro in mammalian cells leads to ER accumulation of ceramides and apoptosis but ceramide levels are not altered in transgenic mice lacking its biosynthetic enzymes
Catalog No. 50-293-4137 Supplier Cayman Chemical Supplier No. 244535 MG
A mixture of ceramide phosphoethanolamines; produced and used by invertebrates such as Drosophila, which lacks sphingomyelin and produced only in very small amounts in mammals; has a role in glial ensheathment of axons in Drosophila; depletion in vitro in mammalian cells leads to ER accumulation of ceramides and apoptosis but ceramide levels are not altered in transgenic mice lacking its biosynthetic enzymes