Learn More
Medchemexpress LLC HY-113679 25mg Medchemexpress, Ceramides Mixture CAS:100403-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11367925MG
Encompass_Preferred
Medchemexpress, HY-113679 25mg Ceramides Mixture CAS:100403-19-8 Ceramides Mixture is an endogenous ceramide and consists of hydroxy and non-hydroxy fatty acid-containing ceramides. Ceramides Mixture is a main lipid component of the permeability barrier in epidermis. Ceramides Mixture is involved in the regulation of growth inhibition, cell cycle arrest, and modulation of telomerase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0864