Learn More
Medchemexpress LLC HY-116147 5mg Medchemexpress, Ceranib-2 CAS:1402830-75-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1161475MG
Encompass_Preferred
Medchemexpress, HY-116147 5mg Ceranib-2 CAS:1402830-75-4 Ceranib-2 is a potent and nonlipid ceramidase inhibitor that inhibits cellular ceramidase activity with an IC50 of 28 μM in SKOV3 cells. Ceranib-2 induces the accumulation of multiple ceramide species, decreases levels of sphingosine and sphingosine-1-phosphate (S1P), and induces cell apoptosis. Anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0887