Learn More
Cayman Chemical D-Pinitol, Purity: ≥95%, CAS: 10284-63-6, 10 g
SDP
Catalog No. 502932969
Encompass_Preferred
A cyclitol with diverse biological activities; decreases NF-κB gene expression and induces DNA fragmentation and apoptosis in MCF-7 breast cancer cells; induces endothelium- and nitric oxide-dependent vasodilation in isolated mouse mesenteric arteries; reduces systolic blood pressure in mice at 10 mg/kg; enhances creatine clearance, reduces tubulointerstitial fibrosis, and increases levels of the cytosolic antioxidant enzymes SOD1 and NQO1 in a mouse model of cyclosporin A-induced nephropathy at 50 mg/kg; decreases plasma glucose levels in a mouse model of streptozotocin-induced diabetes
Catalog No. 50-293-2969 Supplier Cayman Chemical Supplier No. 2681510 G
A cyclitol with diverse biological activities; decreases NF-κB gene expression and induces DNA fragmentation and apoptosis in MCF-7 breast cancer cells; induces endothelium- and nitric oxide-dependent vasodilation in isolated mouse mesenteric arteries; reduces systolic blood pressure in mice at 10 mg/kg; enhances creatine clearance, reduces tubulointerstitial fibrosis, and increases levels of the cytosolic antioxidant enzymes SOD1 and NQO1 in a mouse model of cyclosporin A-induced nephropathy at 50 mg/kg; decreases plasma glucose levels in a mouse model of streptozotocin-induced diabetes