Learn More
Med Vet International Cerclage Wire Passer, 15cm, J0217, Each
Catalog No. 5000585489
Encompass_Preferred
This is a simple instrument for passage of cerclage wire around fracture sites. Stainless steel. 15cm.Minimizes trauma to surrounding tissueSaves surgical timeDouble-ended for a whole range of bone sizes
Catalog No. 50-005-85489 Supplier Med Vet International Supplier No. JOR217
This is a simple instrument for passage of cerclage wire around fracture sites. Stainless steel. 15cm.Minimizes trauma to surrounding tissueSaves surgical timeDouble-ended for a whole range of bone sizes