Learn More
Cayman Chemical CercosporamIde 2.5mg
SDP
Catalog No. 502968598
Encompass_Preferred
A natural antifungal phytotoxin that selectively and potently inhibits fungal Pkc1, which are central to cell wall integrity (IC50 = 25 nM for Candida Pkc1); potently inhibits MAPK-interacting kinases Mnk1 and Mnk2 (IC50 = 115 and 11 nM, respectively), reducing protein translation in cancer cells
Catalog No. 50-296-8598 Supplier Cayman Chemical Supplier No. 155002.5 MG
A natural antifungal phytotoxin that selectively and potently inhibits fungal Pkc1, which are central to cell wall integrity (IC50 = 25 nM for Candida Pkc1); potently inhibits MAPK-interacting kinases Mnk1 and Mnk2 (IC50 = 115 and 11 nM, respectively), reducing protein translation in cancer cells