Learn More
AdipoGen Cercosporin (1 mg)
SDP
Supplier:  AdipoGen AGCN20111M001
Encompass_Preferred
Chemical. CAS: 35082-49-6. Formula: C29H26O10. MW: 534.5. Isolated from Cercospora sp. Phytotoxin. Potent and specific PKC inhibitor. Competes for the phorbol binding site. Anticancer compound. Cytotoxic. Antiproliferative. Antiparasitic. Antimicrobial.
Catalog No. 50-205-3472