Learn More
Abcam Cercosporin, PKC inhibitor, 1MG
Catalog No. 502705981
Encompass_Preferred
MW 534.5 Da, Purity >95%. Light-induced phytotoxin. Potent, selective competitive PKC inhibitor. Shows antiproliferative effects (IC₅₀ = 4.68 μM, MCF7 cancer cell lines). Shows antimicrobial and antiparasitic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5981 Supplier Abcam Supplier No. AB1442181MG
MW 534.5 Da, Purity >95%. Light-induced phytotoxin. Potent, selective competitive PKC inhibitor. Shows antiproliferative effects (IC₅₀ = 4.68 μM, MCF7 cancer cell lines). Shows antimicrobial and antiparasitic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement