Learn More
Cayman Chemical Cercosporin, Purity: ≥95%, CAS: 35082-49-6, 5 mg
SDP
Catalog No. 502986979
Encompass_Preferred
A cytotoxic phytotoxin; produces ROS in plant hosts; has light-activated cytotoxic effects (CC50s = 241, 282, and 174 nM in HeLa, A432, and MCF-7 cells, respectively); a selective inhibitor of PKC (IC50 = 600-1,300 nM) with no activity on PKA or PPK
Catalog No. 50-298-6979 Supplier Cayman Chemical Supplier No. 221755 MG
A cytotoxic phytotoxin; produces ROS in plant hosts; has light-activated cytotoxic effects (CC50s = 241, 282, and 174 nM in HeLa, A432, and MCF-7 cells, respectively); a selective inhibitor of PKC (IC50 = 600-1,300 nM) with no activity on PKA or PPK