Learn More
Medchemexpress LLC HY-15999 10mg Medchemexpress, Cerdulatinib CAS:1198300-79-6 Purity:>98%
SDP
Catalog No. 502029184
Encompass_Preferred
Medchemexpress, HY-15999 10mg Cerdulatinib CAS:1198300-79-6 Cerdulatinib (PRT062070) is a selective Tyk2 inhibitor with an IC50 of 0.5 nM. Cerdulatinib (PRT062070) also is a dual JAK and SYK inhibitor with IC50s of 12, 6, 8 and 32 for JAK1, 2, 3 and SYK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9184 Supplier Medchemexpress LLC Supplier No. HY1599910MG
Medchemexpress, HY-15999 10mg Cerdulatinib CAS:1198300-79-6 Cerdulatinib (PRT062070) is a selective Tyk2 inhibitor with an IC50 of 0.5 nM. Cerdulatinib (PRT062070) also is a dual JAK and SYK inhibitor with IC50s of 12, 6, 8 and 32 for JAK1, 2, 3 and SYK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.