Learn More
TARGETMOL CHEMICALS INC CERDULATINIB HYDROCHLORI 5MG
SDP
Catalog No. 502579557
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cerdulatinib hydrochloride (PRT2070 hydrochloride) is an oral active multi-targeted tyrosine kinase inhibitor with IC50 of 12 nM/6 nM/8 nM/0.5 nM and 32 nM for JAK1/JAK2/JAK3/TYK2 and Syk respectively. Also inhibits 19 other tested kinases with IC50 less than 200 nM. purity: 99%
Catalog No. 50-257-9557 Supplier TARGETMOL CHEMICALS INC Supplier No. T61045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cerdulatinib hydrochloride (PRT2070 hydrochloride) is an oral active multi-targeted tyrosine kinase inhibitor with IC50 of 12 nM/6 nM/8 nM/0.5 nM and 32 nM for JAK1/JAK2/JAK3/TYK2 and Syk respectively. Also inhibits 19 other tested kinases with IC50 less than 200 nM. purity: 99%