Learn More
Medchemexpress LLC Cerdulatinib hydrochloride | 1369761-01-2 | MFCD28348366 | 99.0% | 482.00 g·mol⁻¹ | C20H28ClN7O3S | 10 MG
SDP
Catalog No. 5000285567
Encompass_Preferred
Cerdulatinib hydrochloride is a selective, orally active, reversible ATP-competitive inhibitor of spleen tyrosine kinase (SYK) and Janus kinases (JAKs). It is used as a research compound for studies of immune signaling, autoimmune disease models, and B-cell malignancies.
  • Selective dual SYK and JAK inhibition suitable for signaling studies.
  • High reported purity appropriate for biochemical and cellular assays.
  • Available in small solid quantities for preclinical research use.
  • Soluble in DMSO, enabling preparation of concentrated assay stocks.
Catalog No. 50-002-85567 Supplier Medchemexpress LLC Supplier No. HY15999A10MG
Cerdulatinib hydrochloride is a selective, orally active, reversible ATP-competitive inhibitor of spleen tyrosine kinase (SYK) and Janus kinases (JAKs). It is used as a research compound for studies of immune signaling, autoimmune disease models, and B-cell malignancies.
  • Selective dual SYK and JAK inhibition suitable for signaling studies.
  • High reported purity appropriate for biochemical and cellular assays.
  • Available in small solid quantities for preclinical research use.
  • Soluble in DMSO, enabling preparation of concentrated assay stocks.