Learn More
CPC Scientific H-Ser-Gly-Ser-Ala-Lys-Val-Ala-Phe-Ser-Ala-Ile-Arg-Ser-Thr-Asn-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP109A
Encompass
ONE-LETTER SEQUENCE: SGSAKVAFSAIRSTNHMOLECULAR FORMULA: C69H113N23O23MOLECULAR WEIGHT:1632.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [94071-26-8], RESEARCH AREA: NeuropeptidesREFERENCES: J.R. Slemmon et al., PNAS, 81, 6866 (1984)
Catalog No. 50-210-9941