Learn More
Innovative Research Inc Single Donor Human Cerebrospinal Fluid (CSF) 1ml
Catalog No. 502036082
Encompass_Preferred
Single Donor Human Cerebrospinal Fluid (CSF) from Innovative Research is collected from remnant samples from living individual donors. Cerebrospinal fluid (CSF) is a clear, colorless bodily fluid produced in the choroid plexus of the brain. It acts as a cushion or buffer for the cortex, providing basic mechanical and immunological protection to the brain inside the skull, serving a vital function in cerebral auto-regulation of cerebral blood flow.
Catalog No. 50-203-6082 Supplier Innovative Research Inc Supplier No. IRHUCSFSVIAL
Single Donor Human Cerebrospinal Fluid (CSF) from Innovative Research is collected from remnant samples from living individual donors. Cerebrospinal fluid (CSF) is a clear, colorless bodily fluid produced in the choroid plexus of the brain. It acts as a cushion or buffer for the cortex, providing basic mechanical and immunological protection to the brain inside the skull, serving a vital function in cerebral auto-regulation of cerebral blood flow.