Learn More
ATCC Rhodobacter azotoformans; Strain ATH 2.4.3; genomic DNA
Catalog No. 502382510
Encompass
Genomic DNA isolated from Cereibacter azotoformans strain ATH 2.4.3 (ATCC 17025). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2510 Supplier ATCC Supplier No. 17025D5
Genomic DNA isolated from Cereibacter azotoformans strain ATH 2.4.3 (ATCC 17025). This whole-genome sequenced product can be used in PCR and other molecular biology applications.