Learn More
Speware Corporation CEREX SYSTEM 96 PROCESSOR
Catalog No. NC0700526
Encompass
CEREX SYSTEM 96 PROCESSOR. The new CEREX� System 96 processor for Positive Pressure SPE offers state-of-the-art operation for SPE in 96-well plate formats. Positive pressure processing of SPE plates has numerous advantages compared to traditional vacuum manifold processing. Includes: Manifold Non-Segmented Non-Heated, 96 Deep Well Collection Plate 1.2mL, 96 Deep Well Collection Plate 2.0mL, Single Well Waste Plate for 96-Place Systems, Column Seal for 96-Place Systems, Removable, RCB, Aluminum Holder for 1mL RCH Plates, Processor Installation Kit
Catalog No. NC0700526 Supplier Speware Corporation Supplier No. 2880001
Add to Cart
Add to Cart
Request a Quote
CEREX SYSTEM 96 PROCESSOR. The new CEREX� System 96 processor for Positive Pressure SPE offers state-of-the-art operation for SPE in 96-well plate formats. Positive pressure processing of SPE plates has numerous advantages compared to traditional vacuum manifold processing. Includes: Manifold Non-Segmented Non-Heated, 96 Deep Well Collection Plate 1.2mL, 96 Deep Well Collection Plate 2.0mL, Single Well Waste Plate for 96-Place Systems, Column Seal for 96-Place Systems, Removable, RCB, Aluminum Holder for 1mL RCH Plates, Processor Installation Kit