Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Ammonium Molybdate Solution (contains H2SO4) [for TLC Stain], TCI America™
SDP

Catalog No. C1794500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1794500ML 500 mL
C1794100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1794500ML Supplier TCI America Supplier No. C1794500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD06797081
Synonym CAM Solution

Specifications

Color Yellow
Quantity 500 mL
UN Number 3264
Physical Form Liquid
Chemical Name or Material Ceric Ammonium Molybdate Solution (contains H2SO4) [for TLC Stain]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.