Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Ammonium Nitrate ACS AR, Macron Fine Chemicals™

Catalog No. MK2568500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-2568-500 POLYSTORMOR; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-2568-500 Supplier Avantor Supplier No. 256804

missing translation for 'validMainframeMsg'

Specifications

CAS 16774-21-3
Quantity 500 g
Molecular Formula (NH4)2Ce(NO3)6
Formula Weight 548.23
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Ceric Ammonium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.