Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 1,2-Dinitroglycerin Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0101ML

Catalog No. 01-001-155


Add to Cart

Description

Description

1,2-Dinitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution™ is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 621-65-8
MFCD00661060
1.0 mg/mL in acetonitrile
Ampule
C3H6N2O7
UN 1648 3 / PGII
182.09
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-835-2

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation