Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 1-(3-Chlorophenyl)piperazine-d8 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001075 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-075 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-075 Supplier MilliporeSigma Supelco Supplier No. C1121ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An internal standard suitable for quantitation of 1-(3-Chlorophenyl)piperazine levels in urine or blood by GC/MS or LC-MS/MS. 1-(3-Chlorophenyl)piperazine, also known as mCPP, is a phenyl piperazine derivative and club/designer drug used in combination with Benzylpiperazine (BZP) and Trifluoromethylphenylpiperazine (TFMPP) as an alternative to LSD and MDMA. mCPP is also the primary, active metabolite in urine of the antidepressants trazodone and nefazodone.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1313393-63-3
Linear Formula C10D8H6Cl2N2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym 1-(3-Chlorophenyl)piperazine-D8 hydrochloride solution
Formula Weight 241.19
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.