Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 1,3-Dinitroglycerin Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001153 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-153 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-153 Supplier MilliporeSigma Supelco Supplier No. D0031ML

missing translation for 'validMainframeMsg'

Certified reference material

1,3-Dinitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution™ is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 623-87-0
Linear Formula C3H6N2O7
MDL Number MFCD00661061
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 182.09
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.