Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 10,11-Dihydro-10-Hydroxycarbamazepine Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco D0911ML

Catalog No. 01-001-211


Add to Cart

Description

Description

Also known as monohydroxy carbamazepine (MHC), 10,11-Dihydro-10-Hydroxycarbamazepine is a pharmacologically-active, primary metabolite of oxcarbazepine in urine. Oxcarbazepine, sold under the trade name Trileptal™, is an anticonvulsant drug used primarily for the treatment of epilepsy along with reduction of anxiety symptoms and treatment of mood disorders. Suitable uses for this certified solution standard include as a starting material for calibrators and controls in LC/MS or GC/MS oxcarbazepine testing applications for clinical toxicology, urine drug testing or forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 29331-92-8
MFCD00871817
1.0 mg/mL in acetonitrile
Ampule
2°C (35.6°F)
C15H14N2O2
UN 1648 3 / PGII
254.28
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation