Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 10-Ketonaltrexone Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001519 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-519 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-519 Supplier MilliporeSigma Supelco Supplier No. IMPN007031ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

10-Ketonaltrexone is a potential degradation product of naltrexone hydrochloride. This certified solution standard is appropriate for use in LC/MS or GC/MS applications such as impurity identification in naltrexone formulations. Naltrexone is a pharmaceutical opioid receptor antagonist used in the treatment of alcohol and opioid dependence.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 96445-14-6
MDL Number MFCD08275608
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.