Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-11-nor-9-Carboxy-Delta9-THC-D3 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0081ML

Catalog No. 01-002-033


Add to Cart

Description

Description

11-nor-9-Carboxy-δ9-THC, also known as THC acid, is the main secondary metabolite of δ9-THC found in urine or blood of cannabis users. This internal standard is applicable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug testing and isotope dilution methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 136844-96-7
MFCD00673309
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C21H25O4D3
UN 1230 - class 3 - PG 2 - Methanol, solution
347.46
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation