Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-11-nor-9-Carboxy-Delta9-THC Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0101ML

Catalog No. 01-002-035


Add to Cart

Description

Description

A certified solution standard of (±)-11-nor-9-Carboxy-Δ9-THC at a higher concentration of 1.0 mg/mL. Also known as THC acid, 11-nor-9-Carboxy-Δ9-THC is the main secondary metabolite of THC found in urine or blood of Cannabis users. This reference solution is suitable for use as starting materials in the preparation of calibrators and controls for THC testing by GC/MS or LC/MS in forensic analysis, urine drug testing, or clinical toxicology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 104874-50-2
UN 1230 - class 3 - PG 2 - Methanol, solution
344.44
1 mL
C21H28O4
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation