Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-11-nor-9-Carboxy-Delta9-THC Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002031 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-031 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-031 Supplier MilliporeSigma Supelco Supplier No. T0061ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

11-nor-9-Carboxy-δ9-THC, also known as THC acid, is the main secondary metabolite of δ9-THC found in urine or blood of cannabis users. This certified reference solution is suitable as starting materials in the preparation of calibrators and controls for THC testing by GC/MS or LC/MS in forensic analysis, urine drug testing, or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 104874-50-2
Linear Formula C21H28O4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 344.44
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.