Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+)-11-Nor-Delta9-THC-9-carboxylic acid glucuronide Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002051 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-051 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-051 Supplier MilliporeSigma Supelco Supplier No. T0381ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

11-Nor-9-carboxy-δ9-THC glucuronide is a major urinary metabolite of δ9-THC. This certified Snap-N-Spike™ reference solution is suitable for numerous GC/MS or LC/MS applications from pharmaceutical testing and research to clinical toxicology or forensic analysis. This Certified Reference Material provides added value in allowing calibration of THC acid glucuronide by LC-MS/MS methods without the need of an added hydrolysis step.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1362113-32-3
Linear Formula C27H36O10
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 520.57
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.