Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 17alpha-Ethynylestradiol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001303 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-303 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-303 Supplier MilliporeSigma Supelco Supplier No. E0761ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A new Certified Spiking Solution™ and certified solution standard suitable for LC/MS and GC/MS applications from clinical and diagnostic testing, endocrinology, and clinical chemistry to environmental testing for EPA Method 1698. 17α-Ethynylestradiol is one of the most commonly used medications and can be found in almost all modern formulations of combined oral contraceptive pills.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 57-63-6
Linear Formula C20H24O2
MDL Number MFCD01004605
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 296.40
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.