Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 17alpha-Hydroxyprogesterone-2,3,4-13C3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco H1001ML

Catalog No. 01-001-450


 Restricted Item
Add to Cart

Description

Description

A stable labeled internal standard suitable for quantitation of 17α-Hydroxyprogesterone levels in LC-MS/MS testing assays for newborn screening or endocrinology. For LC-MS/MS assays 13C labeled internal standards can offer certain advantages such as eliminating potential issues of isotopic exchange and minimizing separation of later-eluting analytes from a higher substituted deuterium-labeled internal standard.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1356154-92-1
MFCD18632572
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
13C3C18H30O3
UN 1230 - class 3 - PG 2 - Methanol, solution
333.44
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation