Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 17alpha-Hydroxyprogesterone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001441 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-441 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-441 Supplier MilliporeSigma Supelco Supplier No. H0851ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A calibration material for use in newborn screening assays by LC-MS/MS. This Certified Spiking Solution™ is suitable for many critical applications in clinical and research environments from preparation of controls and system tuning/suitability standards to OEM use in preparation of reagents and assay development.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 68-96-2
Linear Formula C21H30O3
MDL Number MFCD00003659
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 330.46
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.