Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ 17beta-Estradiol-2,3,4-13C3 Solution, , Sold by MilliporeSigma™ Supelco™

Catalog No. 01001300 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-300 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-300 Supplier MilliporeSigma Supelco Supplier No. E0731ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A sex hormone, estradiol is measured in women to assess activity of the ovaries and overall reproductive function. This stable labeled internal standard is suitable for quantitation of estradiol levels in women, men, or children by LC/MS in applications for endocrinology, female health testing or clinical chemistry.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 1261254-48-1
Linear Formula 13C3C15H24O2
MDL Number MFCD15144881
UN Number UN 1648 3 / PGII
Formula Weight 275.36
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.